Assay Details
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081490.2 | NP_001074959.1 | 5 - 6 | 465 | 90 | |
NM_023605.2 | NP_076094.2 | 5 - 6 | 767 | 90 | ||
GenBank mRNA | AB041585.1 | - | 5 - 6 | 736 | 90 | |
AK018482.1 | - | 5 - 6 | 718 | 90 | ||
AK077607.1 | - | 5 - 6 | 428 | 90 | ||
BC020074.1 | - | 5 - 6 | 738 | 3710611 | 90 | |
DV657928.1 | - | 5 - 6 | 453 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
