Assay Details
| Gene Symbol: Mia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019394.3 | NP_062267.2 | 3 - 4 | 470 | 67 | |
| GenBank mRNA | AK135151.1 | - | 2 - 3 | 870 | 67 | |
| AW047492.1 | - | 1 - 2 | 67 | |||
| CK794645.1 | - | 3 - 4 | 464 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085335 | BC009815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085335 | CK794645.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085335 | NM_019394.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171693 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085335 | BC009815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085335 | CK794645.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085335 | NM_019394.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174346 | BC009815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174346 | BE864464.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174346 | CK794645.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174346 | X94322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P174346 | NM_019394.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P415167 | X94322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P415168 | X94322.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25015971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171693 | - | - |
| Whole Mouse Genome G4122A | A_52_P174346 | BC009815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174346 | BE864464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174346 | CK794645.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174346 | X94322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P174346 | NM_019394.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P415168 | X94322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P415167 | X94322.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124443 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116764 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135311 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06441596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06441607 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06121128 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06441600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06441605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06394634 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06441602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25016006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25016010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25016004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25016005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25015956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05901012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06535936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05901014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05901001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25015967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561191 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419608_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561187 | - | - |
| Mouse Genome 430 2.0 Array | 1419608_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419608_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12587_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12587_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023066.mm.1 | TC0700004639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023066.mm.1 | NM_019394.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023066.mm.1 | NM_019394.3 | Refseq |
| Mouse Expression Set 430 | 1419608_a_at | - | - |
| Mouse Expression Set 430 | 1419608_PM_a_at | - | - |
| Murine Genome U74v2 Set | 101453_at | - | - |
| Murine Genome U74v2 Set | 101453_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419608_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419608_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419608_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419608_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5700273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777087 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777087 | - | - |
| mouse-6 v1.1 (Illumina) | scl012587.2_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012587.2_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2880657 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2880657 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012587.2_41-S | - | - |

Mia
Genomic Map