Assay Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025616.3 | NP_079892.1 | 5 - 6 | 382 | 61 | |
| GenBank mRNA | AK008340.1 | - | 5 - 6 | 383 | 61 | |
| AK008382.1 | - | 5 - 6 | 380 | 61 | ||
| AK010832.1 | - | 5 - 6 | 348 | 61 | ||
| AK012977.1 | - | 5 - 6 | 379 | 61 | ||
| AK088074.1 | - | 5 - 6 | 375 | 61 | ||
| AK167473.1 | - | 5 - 6 | 385 | 61 | ||
| AK178698.1 | - | 4 - 5 | 156 | 61 | ||
| AY551342.1 | - | 5 - 6 | 368 | 61 | ||
| BC010303.1 | - | 5 - 6 | 357 | 3597676 | 61 | |
| BC037056.1 | - | 5 - 6 | 384 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC010303.1 transcripts.
Set Membership: |
|

Timm50
Genomic Map