Assay Details
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285435.1 | NP_001272364.1 | 11 - 12 | 1307 | 99 | |
NR_104339.1 | 10 - 11 | 1825 | 99 | |||
NR_104340.1 | 10 - 11 | 1822 | 99 | |||
GenBank mRNA | AK028819.1 | - | 11 - 12 | 1779 | 99 | |
AK029081.1 | - | 10 - 11 | 1763 | 99 | ||
AK154947.1 | - | 7 - 8 | 1412 | 99 | ||
BC034843.1 | - | 9 - 10 | 1466 | 99 | ||
BC058577.1 | - | 6 - 7 | 2398 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
