Assay Details
| Gene Symbol: Dyrk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037957.3 | NP_001033046.1 | 4 - 5 | 629 | 87 | |
| NM_001271370.1 | NP_001258299.1 | 4 - 5 | 582 | 87 | ||
| NM_010092.2 | NP_034222.1 | 4 - 5 | 629 | 87 | ||
| GenBank mRNA | AJ537610.1 | - | 4 - 5 | 582 | 87 | |
| AK148342.1 | - | 4 - 5 | 609 | 87 | ||
| BC019545.1 | - | 4 - 5 | 629 | 4511845 | 87 | |
| Y18280.2 | - | 4 - 5 | 618 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk1b
Genomic Map