Assay Details
| Gene Symbol: Smarcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161853.1 | NP_001155325.1 | 4 - 5 | 690 | 78 | |
| NM_011418.2 | NP_035548.1 | 4 - 5 | 717 | 78 | ||
| GenBank mRNA | AB017343.1 | - | 4 - 5 | 498 | 78 | |
| AB017344.1 | - | 4 - 5 | 471 | 78 | ||
| AB041578.1 | - | 4 - 5 | 672 | 78 | ||
| AJ011739.1 | - | 4 - 5 | 691 | 78 | ||
| AJ011740.1 | - | 4 - 5 | 691 | 78 | ||
| AK075906.1 | - | 4 - 5 | 700 | 78 | ||
| AK145552.1 | - | 4 - 5 | 679 | 78 | ||
| AK147638.1 | - | 1 - 2 | 5051 | 78 | ||
| AK147685.1 | - | 1 - 2 | 5051 | 78 | ||
| AK150169.1 | - | 4 - 5 | 691 | 78 | ||
| AK176985.1 | - | 1 - 2 | 36 | 78 | ||
| AK181638.1 | - | 1 - 2 | 36 | 78 | ||
| BC025163.1 | - | 4 - 5 | 671 | 3154501 | 78 | |
| BC035269.1 | - | 2 - 3 | 2819 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smarcb1
Genomic Map