Assay Details
| Gene Symbol: Aire | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271549.1 | NP_001258478.1 | 11 - 12 | 1484 | 101 | |
| NM_001271550.1 | NP_001258479.1 | 11 - 12 | 1475 | 101 | ||
| NM_001271551.1 | NP_001258480.1 | 11 - 12 | 1472 | 101 | ||
| NM_001271552.1 | NP_001258481.1 | 10 - 11 | 1310 | 101 | ||
| NM_001271553.1 | NP_001258482.1 | 10 - 11 | 1307 | 101 | ||
| NM_001271554.1 | NP_001258483.1 | 10 - 11 | 1298 | 101 | ||
| NM_001271555.1 | NP_001258484.1 | 10 - 11 | 1295 | 101 | ||
| NM_009646.2 | NP_033776.1 | 11 - 12 | 1487 | 101 | ||
| NR_073358.1 | 12 - 13 | 1542 | 101 | |||
| GenBank mRNA | AF079536.1 | - | 11 - 12 | 1457 | 101 | |
| AF128115.1 | - | 11 - 12 | 1454 | 101 | ||
| AF128116.1 | - | 11 - 12 | 1445 | 101 | ||
| AF128117.1 | - | 11 - 12 | 1442 | 101 | ||
| AF128118.1 | - | 10 - 11 | 1280 | 101 | ||
| AF128119.1 | - | 10 - 11 | 1277 | 101 | ||
| AF128120.1 | - | 10 - 11 | 1268 | 101 | ||
| AF128121.1 | - | 10 - 11 | 1265 | 101 | ||
| AF128772.1 | - | 11 - 12 | 1400 | 101 | ||
| AF128773.1 | - | 11 - 12 | 1400 | 101 | ||
| AJ132243.1 | - | 11 - 12 | 1400 | 101 | ||
| AJ243821.1 | - | 11 - 12 | 1400 | 101 | ||
| AK153690.1 | - | 11 - 12 | 2487 | 101 | ||
| BC103510.1 | - | 12 - 13 | 1504 | 40039055 | 101 | |
| BC103511.1 | - | 10 - 11 | 1272 | 40039054 | 101 | |
| BC103518.1 | - | 11 - 12 | 1446 | 40039058 | 101 | |
| EU625343.3 | - | 11 - 12 | 1417 | 101 | ||
| JF267399.1 | - | 11 - 12 | 1417 | 101 | ||
More Information
Set Membership: |
|

Aire
Genomic Map