Assay Details
| Gene Symbol: Aire | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271550.1 | NP_001258479.1 | 6 - 7 | 876 | 72 | |
| NM_001271551.1 | NP_001258480.1 | 6 - 7 | 876 | 72 | ||
| NM_001271554.1 | NP_001258483.1 | 6 - 7 | 876 | 72 | ||
| NM_001271555.1 | NP_001258484.1 | 6 - 7 | 876 | 72 | ||
| NM_001271558.1 | NP_001258487.1 | 6 - 7 | 876 | 72 | ||
| NM_001271559.1 | NP_001258488.1 | 6 - 7 | 876 | 72 | ||
| GenBank mRNA | AF128116.1 | - | 6 - 7 | 846 | 72 | |
| AF128117.1 | - | 6 - 7 | 846 | 72 | ||
| AF128120.1 | - | 6 - 7 | 846 | 72 | ||
| AF128121.1 | - | 6 - 7 | 846 | 72 | ||
| AF128124.1 | - | 6 - 7 | 846 | 72 | ||
| AF128125.1 | - | 6 - 7 | 846 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aire
Genomic Map