Assay Details
| Gene Symbol: Psap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011179.3 | NP_035309.3 | 8 - 9 | 898 | 81 | |
| GenBank mRNA | AK148545.1 | - | 8 - 9 | 814 | 81 | |
| S36200.1 | - | 8 - 9 | 788 | 81 | ||
| S71616.1 | - | 8 - 9 | 828 | 81 | ||
More Information
Set Membership: |
|

Psap
Genomic Map