Assay Details
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183152.3 | NP_898975.2 | 10 - 11 | 1235 | 103 | |
GenBank mRNA | AK031968.1 | - | 10 - 11 | 1236 | 103 | |
BC086684.1 | - | 5 - 6 | 532 | 30679265 | 103 | |
BC099679.1 | - | 10 - 11 | 1204 | 6489647 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
