Assay Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173751.4 | NP_776112.1 | 11 - 12 | 1478 | 74 | |
| GenBank mRNA | AK087956.1 | - | 10 - 11 | 1352 | 74 | |
| AK146075.1 | - | 10 - 11 | 1406 | 74 | ||
| AK169899.1 | - | 10 - 11 | 1404 | 74 | ||
| BC052054.1 | - | 10 - 11 | 1812 | 74 | ||
| BC056459.1 | - | 11 - 12 | 1436 | 5718157 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilvbl
Genomic Map