Assay Details
Gene Symbol: Ilvbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173751.4 | NP_776112.1 | 6 - 7 | 886 | 64 | |
GenBank mRNA | AK030933.1 | - | 3 - 4 | 324 | 64 | |
AK087956.1 | - | 5 - 6 | 760 | 64 | ||
AK133553.1 | - | 5 - 6 | 816 | 64 | ||
AK138031.1 | - | 5 - 6 | 735 | 64 | ||
AK143918.1 | - | 5 - 6 | 785 | 64 | ||
AK146075.1 | - | 5 - 6 | 814 | 64 | ||
AK169899.1 | - | 5 - 6 | 812 | 64 | ||
BC052054.1 | - | 5 - 6 | 1220 | 64 | ||
BC056459.1 | - | 6 - 7 | 844 | 5718157 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
