Assay Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010348.3 | NP_034478.1 | 3 - 4 | 1104 | 72 | |
| NM_146072.4 | NP_666184.2 | 3 - 4 | 1104 | 72 | ||
| GenBank mRNA | AK042635.1 | - | 3 - 4 | 1105 | 72 | |
| AK043542.1 | - | 3 - 4 | 1084 | 72 | ||
| AK043737.1 | - | 3 - 4 | 1068 | 72 | ||
| AK044250.1 | - | 3 - 4 | 1063 | 72 | ||
| AK044420.1 | - | 3 - 4 | 1058 | 72 | ||
| AK046122.1 | - | 3 - 4 | 1111 | 72 | ||
| AK048634.1 | - | 3 - 4 | 1075 | 72 | ||
| AK133667.1 | - | 3 - 4 | 1090 | 72 | ||
| BC031822.1 | - | 2 - 3 | 526 | 4501004 | 72 | |
| BC049275.1 | - | 2 - 3 | 526 | 4501004 | 72 | |
| X66118.1 | - | 3 - 4 | 1090 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik1
Genomic Map