Assay Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010348.3 | NP_034478.1 | 6 - 7 | 1511 | 84 | |
| NM_146072.4 | NP_666184.2 | 6 - 7 | 1511 | 84 | ||
| GenBank mRNA | AK042635.1 | - | 6 - 7 | 1512 | 84 | |
| AK043542.1 | - | 6 - 7 | 1491 | 84 | ||
| AK043737.1 | - | 6 - 7 | 1475 | 84 | ||
| AK044250.1 | - | 6 - 7 | 1470 | 84 | ||
| AK046122.1 | - | 6 - 7 | 1518 | 84 | ||
| AK048634.1 | - | 6 - 7 | 1482 | 84 | ||
| AK133667.1 | - | 6 - 7 | 1497 | 84 | ||
| AK193923.1 | - | 1 - 2 | 88 | 84 | ||
| BC031822.1 | - | 5 - 6 | 933 | 4501004 | 84 | |
| BC049275.1 | - | 5 - 6 | 933 | 4501004 | 84 | |
| X66118.1 | - | 6 - 7 | 1497 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik1
Genomic Map