Assay Details
| Gene Symbol: Idh3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029573.2 | NP_083849.1 | 4 - 5 | 291 | 70 | |
| GenBank mRNA | AK003393.1 | - | 1 - 2 | 184 | 70 | |
| AK010065.1 | - | 4 - 5 | 315 | 70 | ||
| AK032787.1 | - | 4 - 5 | 315 | 70 | ||
| AK047951.1 | - | 4 - 5 | 313 | 70 | ||
| AK117103.1 | - | 4 - 5 | 327 | 70 | ||
| AK146717.1 | - | 1 - 2 | 155 | 70 | ||
| AK150618.1 | - | 4 - 5 | 322 | 70 | ||
| AK151304.1 | - | 4 - 5 | 316 | 70 | ||
| AK152353.1 | - | 4 - 5 | 325 | 70 | ||
| AK153459.1 | - | 4 - 5 | 316 | 70 | ||
| AK158646.1 | - | 4 - 5 | 469 | 70 | ||
| AK159051.1 | - | 4 - 5 | 388 | 70 | ||
| AK159666.1 | - | 4 - 5 | 322 | 70 | ||
| AK160054.1 | - | 4 - 5 | 322 | 70 | ||
| AK164038.1 | - | 5 - 6 | 417 | 70 | ||
| AK167822.1 | - | 4 - 5 | 322 | 70 | ||
| AK168034.1 | - | 4 - 5 | 313 | 70 | ||
| AK168049.1 | - | 4 - 5 | 316 | 70 | ||
| AK168149.1 | - | 4 - 5 | 322 | 70 | ||
| AK169152.1 | - | 4 - 5 | 289 | 70 | ||
| BC034273.1 | - | 4 - 5 | 281 | 4166193 | 70 | |
| BC049956.1 | - | 4 - 5 | 291 | 4212315 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Idh3a
Genomic Map