Assay Details
| Gene Symbol: Elovl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148941.2 | NP_683743.2 | 2 - 3 | 547 | 108 | |
| GenBank mRNA | AF277093.1 | - | 2 - 3 | 343 | 108 | |
| AK029065.1 | - | 2 - 3 | 408 | 108 | ||
| AK030666.1 | - | 2 - 3 | 496 | 108 | ||
| BC037030.1 | - | 2 - 3 | 490 | 4503493 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elovl4
Genomic Map