Assay Details
| Gene Symbol: Mns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008613.3 | NP_032639.1 | 4 - 5 | 611 | 71 | |
| GenBank mRNA | AK013048.1 | - | 4 - 5 | 578 | 71 | |
| AK132772.1 | - | 4 - 5 | 586 | 71 | ||
| BC046624.1 | - | 4 - 5 | 524 | 6314904 | 71 | |
| D14849.1 | - | 4 - 5 | 642 | 71 | ||
More Information
Set Membership: |
|

Mns1
Genomic Map