Assay Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301230.1 | NP_001288159.1 | 3 - 4 | 530 | 72 | |
| NM_001301232.1 | NP_001288161.1 | 2 - 3 | 423 | 72 | ||
| NM_023635.6 | NP_076124.1 | 2 - 3 | 381 | 72 | ||
| GenBank mRNA | AB046693.1 | - | 1 - 2 | 179 | 72 | |
| AB120511.1 | - | 1 - 2 | 157 | 72 | ||
| AB232622.1 | - | 1 - 2 | 157 | 72 | ||
| AF304376.1 | - | 2 - 3 | 336 | 72 | ||
| AK010373.1 | - | 2 - 3 | 299 | 72 | ||
| AK030529.1 | - | 2 - 3 | 328 | 72 | ||
| BC008173.1 | - | 3 - 4 | 487 | 3710213 | 72 | |
| BC009656.1 | - | 2 - 3 | 334 | 3590089 | 72 | |
| CV784419.1 | - | 2 - 3 | 292 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab27a
Genomic Map