Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 13 - 14 | 1632 | 79 | |
| GenBank mRNA | AK141214.1 | - | 13 - 14 | 1644 | 79 | |
| AK146234.1 | - | 12 - 13 | 1584 | 79 | ||
| AK146370.1 | - | 12 - 13 | 1584 | 79 | ||
| AK161013.1 | - | 1 - 2 | 98 | 79 | ||
| AK168180.1 | - | 12 - 13 | 1584 | 79 | ||
| AK168208.1 | - | 13 - 14 | 1667 | 79 | ||
| BC065144.1 | - | 13 - 14 | 1632 | 30627207 | 79 | |
| U58987.1 | - | 13 - 14 | 1667 | 79 | ||
| U60318.1 | - | 12 - 13 | 1586 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mre11a
Genomic Map