Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 15 - 16 | 1911 | 73 | |
| GenBank mRNA | AK141214.1 | - | 15 - 16 | 1923 | 73 | |
| AK146234.1 | - | 14 - 15 | 1863 | 73 | ||
| AK146370.1 | - | 14 - 15 | 1863 | 73 | ||
| AK161013.1 | - | 3 - 4 | 377 | 73 | ||
| AK168180.1 | - | 14 - 15 | 1863 | 73 | ||
| BC065144.1 | - | 15 - 16 | 1911 | 30627207 | 73 | |
| U58987.1 | - | 15 - 16 | 1946 | 73 | ||
| U60318.1 | - | 14 - 15 | 1865 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mre11a
Genomic Map