Assay Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028401.1 | 10 - 11 | 1117 | 103 | ||
| GenBank mRNA | AK012656.1 | - | 8 - 9 | 966 | 103 | |
| AK014920.1 | - | 10 - 11 | 1069 | 103 | ||
| AK050165.1 | - | 10 - 11 | 1073 | 103 | ||
| AK166329.1 | - | 10 - 11 | 1072 | 103 | ||
| AK166633.1 | - | 9 - 10 | 2187 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf1d
Genomic Map