Assay Details
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027261.3 | NP_081537.1 | 5 - 6 | 779 | 147 | |
NM_029248.2 | NP_083524.2 | 5 - 6 | 779 | 147 | ||
NR_028401.1 | 5 - 6 | 779 | 147 | |||
GenBank mRNA | AK012656.1 | - | 3 - 4 | 629 | 146 | |
AK012662.1 | - | 5 - 6 | 731 | 147 | ||
AK014920.1 | - | 5 - 6 | 731 | 147 | ||
AK016109.1 | - | 5 - 6 | 733 | 147 | ||
AK050165.1 | - | 5 - 6 | 735 | 147 | ||
AK161656.1 | - | 5 - 6 | 734 | 147 | ||
AK166329.1 | - | 5 - 6 | 734 | 147 | ||
BC027813.1 | - | 3 - 4 | 567 | 147 | ||
BC056964.1 | - | 4 - 5 | 717 | 147 | ||
BC110660.1 | - | 4 - 5 | 721 | 3158156 | 147 | |
BC117068.1 | - | 4 - 5 | 711 | 40126387 | 147 | |
BC117070.1 | - | 4 - 5 | 711 | 40126389 | 147 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
