Assay Details
| Gene Symbol: Prkcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293650.1 | NP_001280579.1 | 2 - 3 | 239 | 95 | |
| NM_001293651.1 | NP_001280580.1 | 2 - 3 | 243 | 95 | ||
| NM_008925.2 | NP_032951.1 | 2 - 3 | 239 | 95 | ||
| GenBank mRNA | AK137661.1 | - | 2 - 3 | 206 | 95 | |
| AK153931.1 | - | 2 - 3 | 214 | 95 | ||
| AK159705.1 | - | 2 - 3 | 210 | 95 | ||
| BY295975.1 | - | 2 - 3 | 240 | 95 | ||
| CD807078.1 | - | 2 - 3 | 208 | 95 | ||
| U92794.1 | - | 2 - 3 | 192 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcsh
Genomic Map