Assay Details
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 18 - 19 | 2064 | 67 | |
GenBank mRNA | AF237669.1 | - | 18 - 19 | 2023 | 67 | |
AF239661.1 | - | 18 - 19 | 2065 | 67 | ||
AK004876.1 | - | 15 - 16 | 2092 | 67 | ||
AK053550.1 | - | 18 - 19 | 2051 | 67 | ||
AK131140.1 | - | 1570 | 67 | |||
AK134654.1 | - | 2046 | 67 | |||
AK165023.1 | - | 15 - 16 | 2453 | 67 | ||
AK165746.1 | - | 16 - 17 | 2357 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26349892 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2086825 | AK131140.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2086825 | AK134654.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2086825 | AF239661.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2086825 | NM_023622.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751697 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425932 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451665 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455637 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821954 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484107 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952934 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823994 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74623 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213011 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82051 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127761 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130692 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2086825 | AK131140.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2086825 | AK134654.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2086825 | AF239661.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2086825 | NM_023622.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349894 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06720164 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130692 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06720119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06720153 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127761 | - | - |
Whole Mouse Genome G4122A | A_52_P440599 | AK165023.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440599 | AK131140.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440599 | AK053550.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440599 | AK134654.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440599 | AF239661.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139034 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133292 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127761 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130692 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903859 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903823 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903821 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903826 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903848 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07903833 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26349892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07831701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07884440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349833 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07583711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07903826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07837781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349827 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07916824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07728472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08055994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07903845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07643425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08046990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349899 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10591685 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591669 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591671 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591674 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591689 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591675 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591678 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591676 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591670 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591687 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591688 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591679 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591677 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591686 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591672 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591680 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591690 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591673 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591682 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591683 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591681 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431829_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10591668 | - | - |
Mouse Genome 430 2.0 Array | 1431829_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1431829_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71746_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71746_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900015924.mm.1 | NM_023622.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900015924.mm.1 | NM_023622.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900015924.mm.1 | TC0900001890.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900015925.mm.1 | NM_023622.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900015925.mm.1 | NM_023622.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900015925.mm.1 | TC0900001890.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431829_PM_a_at | - | - |
Mouse Expression Set 430 | 1431829_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1431829_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1431829_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431829_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431829_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6550025 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006550025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239492 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1239492 | - | - |
mouse-6 v1.1 (Illumina) | scl36127.13.1_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6550025 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36127.13.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550025 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1239492 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2991779 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239492 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2991779 | - | - |
