Assay Details
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 6 - 7 | 839 | 96 | |
GenBank mRNA | AF237669.1 | - | 6 - 7 | 798 | 96 | |
AF239661.1 | - | 6 - 7 | 840 | 96 | ||
AK004876.1 | - | 5 - 6 | 572 | 96 | ||
AK053550.1 | - | 6 - 7 | 826 | 96 | ||
AK134654.1 | - | 821 | 96 | |||
AK165023.1 | - | 6 - 7 | 827 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
