Assay Details
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 7 - 8 | 1053 | 75 | |
GenBank mRNA | AF237669.1 | - | 7 - 8 | 1012 | 75 | |
AF239661.1 | - | 7 - 8 | 1054 | 75 | ||
AK004876.1 | - | 6 - 7 | 786 | 75 | ||
AK053550.1 | - | 7 - 8 | 1040 | 75 | ||
AK131140.1 | - | 559 | 75 | |||
AK134654.1 | - | 1035 | 75 | |||
AK165023.1 | - | 7 - 8 | 1041 | 75 | ||
AK165746.1 | - | 7 - 8 | 1047 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
