Assay Details
Gene Symbol: Car12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052639.1 | - | 5 - 6 | 658 | 106 | |
AK157977.1 | - | 5 - 6 | 658 | 106 | ||
AK166132.1 | - | 5 - 6 | 656 | 106 | ||
AK170859.1 | - | 5 - 6 | 617 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
