Assay Details
| Gene Symbol: Arhgap42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027823.1 | NP_082099.1 | 11 - 12 | 1297 | 100 | |
| GenBank mRNA | AK082857.1 | - | 11 - 12 | 1385 | 100 | |
| BC137891.1 | - | 11 - 12 | 1297 | 8860910 | 100 | |
| BC137892.1 | - | 11 - 12 | 1297 | 8860911 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap42
Genomic Map