Assay Details
| Gene Symbol: Arhgap42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027823.1 | NP_082099.1 | 13 - 14 | 1437 | 77 | |
| GenBank mRNA | AK082857.1 | - | 13 - 14 | 1525 | 77 | |
| BC137891.1 | - | 13 - 14 | 1437 | 8860910 | 77 | |
| BC137892.1 | - | 13 - 14 | 1437 | 8860911 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap42
Genomic Map