Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011753.3 | NP_035883.2 | 3 - 4 | 483 | 115 | |
| GenBank mRNA | AK017201.1 | - | 3 - 4 | 313 | 115 | |
| AK029298.1 | - | 3 - 4 | 308 | 115 | ||
| AK082002.1 | - | 3 - 4 | 317 | 115 | ||
| AK085739.1 | - | 3 - 4 | 411 | 115 | ||
| AK141740.1 | - | 3 - 4 | 320 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029298.1, AK082002.1, AK141740.1, NM_011753.3 transcripts.
Set Membership: |
|

Zfp26
Genomic Map