Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900015293.mm.1 |
NM_011753.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015293.mm.1 |
NM_011753.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015293.mm.1 |
TC0900001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015298.mm.1 |
NM_011753.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015298.mm.1 |
NM_011753.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015298.mm.1 |
TC0900001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015297.mm.1 |
NM_011753.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015297.mm.1 |
NM_011753.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015297.mm.1 |
TC0900001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015299.mm.1 |
NM_011753.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015299.mm.1 |
NM_011753.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015299.mm.1 |
TC0900001836.mm.1 |
MTA TC |