Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011753.3 | NP_035883.2 | 4 - 5 | 582 | 72 | |
| GenBank mRNA | AK082002.1 | - | 4 - 5 | 416 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp26
Genomic Map