Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011753.3 | NP_035883.2 | 5 - 6 | 684 | 74 | |
| GenBank mRNA | AK082002.1 | - | 5 - 6 | 518 | 74 | |
| AK141740.1 | - | 4 - 5 | 1047 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141740.1 transcripts.
Set Membership: |
|

Zfp26
Genomic Map