Assay Details
Gene Symbol: Zfp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011753.3 | NP_035883.2 | 6 - 7 | 770 | 63 | |
GenBank mRNA | AK082002.1 | - | 6 - 7 | 604 | 63 | |
AK141740.1 | - | 5 - 6 | 1133 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK141740.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
