Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011753.3 | NP_035883.2 | 7 - 8 | 901 | 75 | |
| GenBank mRNA | AK029298.1 | - | 5 - 6 | 1166 | 75 | |
| AK082002.1 | - | 7 - 8 | 735 | 75 | ||
| AK141740.1 | - | 6 - 7 | 1264 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029298.1 transcripts.
Set Membership: |
|

Zfp26
Genomic Map