Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023138.4 | NP_075627.2 | 2 - 3 | 502 | 59 | |
| GenBank mRNA | AK009392.1 | - | 2 - 3 | 519 | 59 | |
| AK080574.1 | - | 2 - 3 | 503 | 59 | ||
| AK138130.1 | - | 2 - 3 | 478 | 59 | ||
| AK143425.1 | - | 2 - 3 | 482 | 59 | ||
| AK171584.1 | - | 2 - 3 | 494 | 59 | ||
| AK172055.1 | - | 2 - 3 | 503 | 59 | ||
| BC014830.1 | - | 2 - 3 | 484 | 4483473 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map2k2
Genomic Map