Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023138.4 | NP_075627.2 | 6 - 7 | 904 | 84 | |
| GenBank mRNA | AK009392.1 | - | 6 - 7 | 921 | 84 | |
| AK080574.1 | - | 6 - 7 | 905 | 84 | ||
| AK138130.1 | - | 6 - 7 | 1013 | 84 | ||
| AK171584.1 | - | 6 - 7 | 896 | 84 | ||
| AK172055.1 | - | 6 - 7 | 905 | 84 | ||
| BC014830.1 | - | 6 - 7 | 886 | 4483473 | 84 | |
| S68267.1 | - | 6 - 7 | 825 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k2
Genomic Map