Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011247.1 | - | 7 - 8 | 1119 | 111 | |
| AK138130.1 | - | 7 - 8 | 1232 | 111 | ||
| AK171584.1 | - | 7 - 8 | 1114 | 111 | ||
| AK172055.1 | - | 7 - 8 | 1123 | 111 | ||
| BC014830.1 | - | 7 - 8 | 1104 | 4483473 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map2k2
Genomic Map