Assay Details
Gene Symbol: Raver1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027911.3 | NP_082187.1 | 2 - 3 | 293 | 64 | |
GenBank mRNA | AK039928.1 | - | 2 - 3 | 305 | 64 | |
AK051444.1 | - | 2 - 3 | 305 | 64 | ||
AK079191.1 | - | 2 - 3 | 298 | 64 | ||
AK084478.1 | - | 2 - 3 | 305 | 64 | ||
AK138596.1 | - | 2 - 3 | 305 | 64 | ||
AK168436.1 | - | 2 - 3 | 314 | 64 | ||
BC058271.1 | - | 2 - 3 | 293 | 6490833 | 64 | |
BC108392.1 | - | 2 - 3 | 292 | 6393833 | 64 |
Gene Symbol: Gm38431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038081.2 | 2 - 3 | 313 | 64 | ||
GenBank mRNA | AK004919.1 | - | 2 - 3 | 306 | 64 | |
AY275472.1 | - | 2 - 3 | 292 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
