Assay Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026545.3 | NP_080821.3 | 4 - 5 | 738 | 83 | |
GenBank mRNA | AK002921.1 | - | 4 - 5 | 577 | 83 | |
AK003436.1 | - | 4 - 5 | 579 | 83 | ||
AK027973.1 | - | 4 - 5 | 542 | 83 | ||
AK051257.1 | - | 4 - 5 | 590 | 83 | ||
AK150493.1 | - | 4 - 5 | 581 | 83 | ||
AK150982.1 | - | 4 - 5 | 545 | 83 | ||
AK153329.1 | - | 4 - 5 | 579 | 83 | ||
AK159216.1 | - | 4 - 5 | 547 | 83 | ||
AK159795.1 | - | 4 - 5 | 563 | 83 | ||
AK159926.1 | - | 4 - 5 | 573 | 83 | ||
AK166718.1 | - | 4 - 5 | 572 | 83 | ||
AK167948.1 | - | 4 - 5 | 547 | 83 | ||
AK168886.1 | - | 4 - 5 | 546 | 83 | ||
BC004075.1 | - | 4 - 5 | 556 | 3591796 | 83 | |
BC005717.1 | - | 4 - 5 | 466 | 2609781 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
