Assay Details
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029364.3 | NP_083640.1 | 3 - 4 | 538 | 115 | |
GenBank mRNA | AK030773.1 | - | 3 - 4 | 537 | 115 | |
AK049162.1 | - | 3 - 4 | 539 | 115 | ||
AK054046.1 | - | 3 - 4 | 538 | 115 | ||
AK083597.1 | - | 3 - 4 | 516 | 115 | ||
AK149863.1 | - | 3 - 4 | 523 | 115 | ||
AK151037.1 | - | 1 - 2 | 160 | 115 | ||
AK159562.1 | - | 3 - 4 | 505 | 115 | ||
AK165180.1 | - | 3 - 4 | 518 | 115 | ||
AK165917.1 | - | 3 - 4 | 488 | 115 | ||
AK169485.1 | - | 3 - 4 | 520 | 115 | ||
AK170791.1 | - | 3 - 4 | 534 | 115 | ||
BC055328.1 | - | 3 - 4 | 532 | 6400020 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
