Assay Details
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029364.3 | NP_083640.1 | 4 - 5 | 607 | 70 | |
GenBank mRNA | AK030773.1 | - | 4 - 5 | 606 | 70 | |
AK049162.1 | - | 4 - 5 | 608 | 70 | ||
AK054046.1 | - | 4 - 5 | 607 | 70 | ||
AK083597.1 | - | 4 - 5 | 585 | 70 | ||
AK149863.1 | - | 4 - 5 | 592 | 70 | ||
AK151037.1 | - | 2 - 3 | 229 | 70 | ||
AK159562.1 | - | 4 - 5 | 574 | 70 | ||
AK165180.1 | - | 4 - 5 | 587 | 70 | ||
AK165917.1 | - | 4 - 5 | 557 | 70 | ||
AK169485.1 | - | 4 - 5 | 589 | 70 | ||
AK170791.1 | - | 4 - 5 | 603 | 70 | ||
BC055328.1 | - | 4 - 5 | 601 | 6400020 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
