Assay Details
| Gene Symbol: Msrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177092.4 | NP_796066.1 | 4 - 5 | 437 | 107 | |
| GenBank mRNA | AK086975.1 | - | 4 - 5 | 404 | 107 | |
| AK137163.1 | - | 4 - 5 | 404 | 107 | ||
| BB617432.1 | - | 4 - 5 | 472 | 107 | ||
| BC096447.1 | - | 4 - 5 | 318 | 6310686 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msrb3
Genomic Map