Assay Details
Gene Symbol: Wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201370.2 | NP_958758.2 | 8 - 9 | 1490 | 84 | |
GenBank mRNA | AK139573.1 | - | 8 - 9 | 1449 | 84 | |
AK143336.1 | - | 8 - 9 | 1414 | 84 | ||
AK163344.1 | - | 8 - 9 | 1492 | 84 | ||
BC052883.1 | - | 8 - 9 | 1397 | 30053490 | 84 | |
BC080784.1 | - | 8 - 9 | 1415 | 30481521 | 84 | |
DQ011691.1 | - | 8 - 9 | 1311 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
