Assay Details
Gene Symbol: Capza2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007604.2 | NP_031630.1 | 7 - 8 | 606 | 120 | |
GenBank mRNA | AK007550.1 | - | 7 - 8 | 605 | 120 | |
AK008204.1 | - | 7 - 8 | 592 | 120 | ||
AK150751.1 | - | 7 - 8 | 673 | 120 | ||
AK152674.1 | - | 4 - 5 | 400 | 120 | ||
AY544984.1 | - | 7 - 8 | 602 | 120 | ||
BC082589.1 | - | 7 - 8 | 606 | 30626416 | 120 | |
U16741.1 | - | 7 - 8 | 687 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
