Assay Details
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144144.1 | - | 12 - 13 | 2007 | 96 | |
BC145267.1 | - | 11 - 12 | 1800 | 9053821 | 96 | |
BC150745.1 | - | 11 - 12 | 1800 | 9087656 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
