Assay Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 17 - 18 | 1939 | 83 | |
| GenBank mRNA | AK154778.1 | - | 17 - 18 | 1851 | 83 | |
| AY083469.1 | - | 17 - 18 | 1601 | 83 | ||
| BC150156.1 | - | 17 - 18 | 1604 | 83 | ||
| BC150478.1 | - | 17 - 18 | 1604 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka3
Genomic Map