Assay Details
Gene Symbol: Prl5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023746.4 | NP_076235.1 | 2 - 3 | 273 | 132 | |
GenBank mRNA | AF226611.1 | - | 2 - 3 | 220 | 132 | |
AK005447.1 | - | 2 - 3 | 274 | 132 | ||
BC051672.1 | - | 2 - 3 | 260 | 30017843 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
