Assay Details
Gene Symbol: Prl5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023746.4 | NP_076235.1 | 4 - 5 | 564 | 152 | |
GenBank mRNA | AF226611.1 | - | 4 - 5 | 511 | 152 | |
AK005447.1 | - | 4 - 5 | 565 | 152 | ||
BC051672.1 | - | 4 - 5 | 551 | 30017843 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | AF226611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | AK005447.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | BC051672.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | AF226611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | AK005447.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | BC051672.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181933 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292337 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889029 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114964 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121080 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | AF226611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | AK005447.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | BC051672.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | AF226611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | AK005447.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | BC051672.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904703 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904707 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904692 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904709 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904690 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904701 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904699 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904694 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121080 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02652681 | - | - |
Whole Mouse Genome G4122A | A_51_P320606 | AF226611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | AK005447.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | BC051672.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | AF226611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | AK005447.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | BC051672.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P320606 | NM_023746.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138620 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120458 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121080 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11030316 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10688085 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10688090 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10688081 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10688087 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10688088 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11030315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904719 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10688081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10688088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904734 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11073091 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904706 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404349 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404348 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404353 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404352 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404351 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404350 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422657_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404347 | - | - |
Mouse Genome 430 2.0 Array | 1422657_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422657_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 28078_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 28078_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001922.mm.1 | NM_023746.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001922.mm.1 | NM_023746.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001922.mm.1 | TC1300000309.mm.1 | MTA TC |
Mouse Expression Set 430 | 1422657_at | - | - |
Mouse Expression Set 430 | 1422657_PM_at | - | - |
Murine Genome U74v2 Set | 168707_PM_i_at | - | - |
Murine Genome U74v2 Set | 168707_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1422657_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422657_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422657_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422657_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 655380 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4480528 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4480528 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004480528 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2676207 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2676207 | - | - |
mouse-6 v1.1 (Illumina) | scl44960.4.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4480528 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44960.4.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480528 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2676207 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2896505 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2676207 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896505 | - | - |
