Assay Details
| Gene Symbol: Slc39a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139143.3 | NP_631882.2 | 7 - 8 | 2415 | 73 | |
| GenBank mRNA | AB071697.1 | - | 6 - 7 | 1798 | 73 | |
| AK028976.1 | - | 7 - 8 | 2374 | 73 | ||
| BC054780.1 | - | 6 - 7 | 1328 | 6813896 | 73 | |
| BC055012.1 | - | 7 - 8 | 2085 | 6337369 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a6
Genomic Map